missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathelicidin, Synthetic Peptide, Invitrogen™
Synthetic Peptide
Brand: Invitrogen GSC00002P
Code : LD
Additional Details : Weight : 0.01000kg
Description
Human CAP-18 Synthetic Peptide, GSC00002P, from Invitrogen.
GSC00002P is the immunizing peptide for the rabbit antibody to human CAP-18 (OSC00009W,OSC00028G)GSC00002P may be used as a control in various immunology applications.Reconstitute with PBS or distilled water.Specifications
820 | |
purified | |
Human Cathelicidin | |
Synthetic Peptide | |
peptide with no preservative | |
CAMP, CAP18, FALL39, FALL-39, 18 kDa cationic antimicrobial protein, CAP-18, hCAP-18, LL37, CRAMP, HSD26 | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P49913 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized | |
Control | |
CAMP | |
No preservative | |
50 μg |