Learn More
Invitrogen™ eBioscience™ Human C3a Platinum ELISA Kit
For the quantitative detection of human C3a.
Brand:  Invitrogen™ BMS2089TWO
Code : Z2
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15309555
  • £478.32 / Pack of 2
Estimated Shipment
03-04-2018
Add to basket
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human C3a
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
Biotin
2 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
complement component 3, ASP, C3a, C3b, C3, Acylation stimulating protein
ELISA
0.07ng/mL
10μL for plasma, serum (prediluted 1:500 for serum, 1:30 for plasma), 50μL for CCS
C3a