missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human C-Reactive Protein/CRP Quantikine ELISA Kit, R&D Systems™
Human C-Reactive Protein/CRP Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 0.78 - 50ng/mL |
---|---|
Assay Sensitivity | 0.022ng/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
Description
The Human C-Reactive Protein/CRP Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human C-Reactive Protein/CRP in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).Specifications
0.78 - 50ng/mL | |
0.022ng/mL | |
Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |