missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human C-Reactive Protein/CRP Quantikine ELISA Kit, R&D Systems™

Human C-Reactive Protein/CRP Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SCRP00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13432696

  • £3544.17 / Pack of 6


Estimated Shipment date
from Supplier 31-03-2020
Add to basket

Description

Description

The Human C-Reactive Protein/CRP Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human C-Reactive Protein/CRP in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Specifications

Specifications

1 Pack
Human
HRP
ELISA
6 Plates
Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Sufficient materials to run ELISAs on at least six 96-well plates
Serum
0.78 - 50ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
1401
0.022ng/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Videos
Documents