missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human BMP-2 DuoSet, R&D Systems™
Human BMP-2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 46.90 to 3000 pg/mL |
---|---|
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gene ID (Entrez) | 650 |
Product Type | ELISA Kit |
Format | 15 96-well Plates |
Description
The Human BMP-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human BMP-2.Specifications
46.90 to 3000 pg/mL | |
650 | |
15 96-well Plates | |
Human |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Kit | |
Biotin |