Learn More
R&D Systems™ Human Free BDNF Quantikine ELISA Kit, R&D Systems™
a 3.5 hour solid phase ELISA designed to measure human BDNF in cell culture supernates, serum, and plasma.
Brand:  R&D Systems™ DBD00
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13462585
  • £572.02 / 96 tests
Estimated Shipment
07-03-2019
Add to basket
Description
Description
It contains recombinant human BDNF expressed in Sf 21 cells and antibodies raised against the recombinant factor. This immunoassay has been shown to quantitate the recombinant BDNF accurately. Results obtained measuring natural human BDNF showed dose-response curves that were parallel to the standard curves obtained using the recombinant Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural human BDNF
Specifications
Specifications
62.50 - 4000pg/mL
<50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules.
627
1 Plate
20pg/mL
Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (50μL), Platelet-poor Heparin Plasma (50μL), Citrate Plasma (50μL)
Sufficient materials to run ELISAs on at least one 96-well plate
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (50uL), Serum (10uL), EDTA Plasma (50uL), Platelet-poor Heparin Plasma (50uL), Citrate Plasma (50uL)