missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Free BDNF Quantikine ELISA Kit, R&D Systems™
Human Free BDNF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 62.50 - 4000pg/mL |
---|---|
Assay Sensitivity | 20pg/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate), Platelet-poor Plasma (Heparin), Serum |
Description
The Human Free BDNF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human BDNF in cell culture supernates (50 ul), serum (10 ul), edta plasma (50 ul), platelet-poor heparin plasma (50 ul), citrate plasma (50 ul).Specifications
62.50 - 4000pg/mL | |
20pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate), Platelet-poor Plasma (Heparin), Serum |