Learn More
Abnova™ Human BDNF (P23560) Recombinant Protein
Used for Func, SDS-PAGE
Brand: Abnova™ P4375.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the nerve growth factor family. It is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of this gene is reduced in both Alzheimer's and Huntington disease patients. This gene may play a role in the regulation of stress response and in the biology of mood disorders. Multiple transcript variants encoding distinct isoforms have been described for this gene. [provided by RefSeq]
Sequence: MHSDPARRGELSVCDSISEWVTAADKKTAVDMSGGTVTVLEKVPVSKGQLKQYFYETKCNPMGYTKEGCRGIDKRHWNSQCRTTQSYVRALTMDSKKRIGWRFIRIDTSCVCTLTIKRGRSpecifications
P23560 | |
Lyophilized | |
27kDa | |
Escherichia coli expression system | |
10 ug | |
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
<0.1 EU/μg | |
BDNF | |
The activity is determined by the dose-dependent induction of C6 cell proliferation. The expected ED50 for this effect is 1.3-2μg/mL. | |
Recombinant | |
Escherichia coli expression system |
Functional Study, SDS-PAGE | |
627 | |
BDNF (Human) Recombinant Protein | |
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue | |
MHSDPARRGELSVCDSISEWVTAADKKTAVDMSGGTVTVLEKVPVSKGQLKQYFYETKCNPMGYTKEGCRGIDKRHWNSQCRTTQSYVRALTMDSKKRIGWRFIRIDTSCVCTLTIKRGR | |
RUO | |
MGC34632 | |
BDNF | |
E. coli | |
None | |
Lyophilized |