missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BACH1 Partial ORF (NP_996749, 396 a.a. - 492 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000571-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16003655

  • £275.34 / 10µg

Estimated Shipment
05-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: HLAKGFWSDICSTDTPCQMQLSPAVAKDGSEQISQKRSECPWLGIRISESPEPGQRTFTTLSSVNCPFISTLSTEGCSSNLEIGNDDYVSEPQQEPC
Specifications

Specifications

NP_996749
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HLAKGFWSDICSTDTPCQMQLSPAVAKDGSEQISQKRSECPWLGIRISESPEPGQRTFTTLSSVNCPFISTLSTEGCSSNLEIGNDDYVSEPQQEPC
RUO
BACH1
Yes
wheat germ expression system
Liquid
571
BACH1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BACH1
Wheat Germ (in vitro)
GST
Videos
Documents