Learn More
Invitrogen™ APRIL Human ELISA Kit
For the quantitative detection of human APRIL.
£451.00 - £3070.00
Specifications
Accession Number | O75888 |
---|---|
Assay Range | 0.78-50.0 ng/mL |
Assay Sensitivity | 0.4 ng/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Includes: Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human APRIL
Biotin-Conjugate anti-human APRIL polyclonal antibody
Streptavidin-HRP
Human APRIL Standard lyophilized, 100ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12737676
|
Invitrogen™
BMS2008 |
96 Tests |
£451.00
Each |
Estimated Shipment: 25-04-2024
Log in to see stock. |
|||||
15591477
|
Invitrogen™
BMS2008TEN |
10 x 96 Tests |
£3070.00
Pack of 10 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
Description
The Human APRIL (Hu A PRoliferation-Inducing Ligand)) ELISA quantitates Hu APRIL in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu APRIL. Principle of the method The Human APRIL solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
O75888 | |
0.4 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
APRIL,CD256,TALL-2,TALL2,TNLG7B,TRDL-1,UNQ383/PRO715,ZTNF2 | |
8.1% | |
HRP | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL | |
TNF 13, TALL 2, TRDL 1, CD256 antigen | |
1 hr. 20 min. |
0.78-50.0 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
8741 | |
7.1% | |
Pre-coated 96 well plate, Adhesive Plate Covers, Sample Diluent, Sample Diluent, Biotinylated Detection Antibody, SAV-HRP, SAV-HRP, Lysis Buffer, Chromogen, Stop Solution, Stop Solution | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 10 min. |
For Research Use Only