missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APRIL Human ELISA Kit
For the quantitative detection of human APRIL.
Brand: Invitrogen™ BMS2008TEN
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human APRIL
Biotin-Conjugate anti-human APRIL polyclonal antibody
Streptavidin-HRP
Human APRIL Standard lyophilized, 100ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Description
Cytokine that binds to TNFRSF13B/TACI and to TNFRSF17/BCMA. Plays a role in the regulation of tumor cell growth. May be involved in monocyte/macrophage-mediated immunological processes. {ECO:0000269; PubMed:10973284}.Specifications
O75888 | |
Biotin | |
8741 | |
ELISA Kit | |
0.4 ng/mL | |
Plasma, 50 μL|Serum, 50 μL|Supernatant, 50 μL | |
1 hr. 20 min. | |
Human | |
TNF 13, TALL 2, TRDL 1, CD256 antigen | |
8.1% | |
Human | |
HRP |
0.78-50.0 ng/mL | |
ELISA | |
Pre-coated 96 well plate, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Lysis Buffer, Chromogen, Stop Solution | |
RUO | |
50 μL | |
2°C to 8°C | |
3 hr. 10 min. | |
APRIL,CD256,TALL-2,TALL2,TNLG7B,TRDL-1,UNQ383/PRO715,ZTNF2 | |
7.1% | |
10 x 96 Tests | |
Supernatant | |
Colorimetric Microplate Reader |
For Research Use Only