missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human anti-Annexin V Platinum ELISA Kit

For the quantitative detection of antibodies to human Annexin V (Vac- alpha).

Brand:  Invitrogen™ BMS247TEN

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15551827

  • £2102.54 / Pack of 10


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with recombinant Annexin V
HRP-Conjugate recombinant Annexin V
Human anti-Annexin V Standard lyophilized, 800ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
10 x 96 tests
Cell culture supernatant, serum
Human
anti Annexin A5, anti Annexin 5
ELISA
1.2ng/mL
50μL
Annexin V
Videos

For Research Use Only