Learn More
Invitrogen™ Angiostatin (Plasminogen) Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHANGIOSTATIN
Code : NEW
Additional Details : Weight : 0.00400kg
Description
The Human Angiostatin (Plasminogen) ELISA quantitates Hu Angiostatin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Angiostatin. Principle of the method The Human Angiostatin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Plasmin dissolves the fibrin of blood clots and acts as a proteolytic factor in a variety of other processes including embryonic development, tissue remodeling, tumor invasion, and inflammation. In ovulation, weakens the walls of the Graafian follicle. It activates the urokinase-type plasminogen activator, collagenases and several complement zymogens, such as C1 and C5. Cleavage of fibronectin and laminin leads to cell detachment and apoptosis. Also cleaves fibrin, thrombospondin and von Willebrand factor. Its role in tissue remodeling and tumor invasion may be modulated by CSPG4. Binds to cells.Specifications
P00747 | |
20 ng/mL | |
ELISA Kit | |
Human | |
5340 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Plasma, 33 μL; Serum, 33 μL; Supernatant, 100 μL | |
Human | |
4 hr. 45 min. |
31.25-2000 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Colorimetric Microplate Reader | |
PLG | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |