missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Angiopoietin-like Protein 7/ANGPTL7 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 10218 |
Molecular Weight (g/mol) | 38.6 kDa |
Name | Angiopoietin-like Protein 7/ANGPTL7 |
Quantity | 25μg |
Description
The Recombinant Human Angiopoietin-like 7 Protein is derived from CHO. The Recombinant Human Angiopoietin-like 7 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
38.6 kDa | |
25μg | |
Chinese Hamster Ovary cell line,CHO-derived human Angiopoietin-like Protein 7/ANGPTL7 protein Met1-Pro346,with a C-terminal 10-His tag |
10218 | |
Angiopoietin-like Protein 7/ANGPTL7 | |
Yes |