missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ALOX12P2 Full-length ORF (AAH41851.1, 1 a.a. - 256 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000245-P01.10ug

 View more versions of this product

Product Code. 16101511

  • £280.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Sequence: MVKLRKHNVLLSLDWFCKWISVQGPGTQGAAFFPCYRWVQGHGIICLPEGTARTVSDDPQNLFKKYREQELEERRWGSWKDGLILPIAGNRQPDLPRDERFLEDKDLDFNVSLAKGLKDLAIKGTLDFINCVKRLEDFKKIFPHGKTVLAERVYDSWKNDAFFGYQFLNGANPMLLRCTSRLPACLVLPPGMEDLKTQLEKELQAGSLFEVDFSLLDGVKPNVIIFKQQCVAAPLVMLKLQPDGGLLPMVIQLQPP
Specifications

Specifications

AAH41851.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ALOX12P2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
245
ALOX12P2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ALOX12E/MGC43625
ALOX12P2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers