missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ACSS2 Partial ORF (NP_061147.1, 32 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055902-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16015585

  • £425.00 / 25µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a cytosolic enzyme that catalyzes the activation of acetate for use in lipid synthesis and energy generation. The protein acts as a monomer and produces acetyl-CoA from acetate in a reaction that requires ATP. Expression of this gene is regulated by sterol regulatory element-binding proteins, transcription factors that activate genes required for the synthesis of cholesterol and unsaturated fatty acids. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: PPEVSRSAHVPSLQRYRELHRRSVEEPREFWGDIAKEFYWKTPCPGPFLRYNFDVTKGKIFIEWMKGATTNICYNVLDRNVHEKKLGDKVAFYWEGNEP
Specifications

Specifications

NP_061147.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PPEVSRSAHVPSLQRYRELHRRSVEEPREFWGDIAKEFYWKTPCPGPFLRYNFDVTKGKIFIEWMKGATTNICYNVLDRNVHEKKLGDKVAFYWEGNEP
RUO
ACSS2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
55902
ACSS2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACAS2/ACS/ACSA/AceCS/DKFZp762G026/dJ1161H23.1
ACSS2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers