Learn More
Abnova™ Human ABHD11 Full-length ORF (NP_683710.1, 1 a.a. - 315 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00083451-P01.2ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein containing an alpha/beta hydrolase fold domain. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]
Sequence: MRAGQQLASMLRWTRAWRLPREGLGPHGPSFARVPVAPSSSSGGRGGAEPRPLPLSYRLLDGEAALPAVVFLHGLFGSKTNFNSIAKILAQQTGRRVLTVDARNHGDSPHSPDMSYEIMSQDLQDLLPQLGLVPCVVVGHSMGGKTAMLLALQRPELVERLIAVDISPVESTGVSHFATYVAAMRAINIADELPRSRARKLADEQLSSVIQDMAVRQHLLTNLVEVDGRFVWRVNLDALTQHLDKILAFPQRQESYLGPTLFLLGGNSQFVHPSHHPEIMRLFPRAQMQTVPNAGHWIHADRPQDFIAAIRGFLVSpecifications
NP_683710.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
RUO | |
ABHD11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
83451 | |
ABHD11 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PP1226/WBSCR21 | |
ABHD11 | |
Recombinant | |
wheat germ expression system |