Learn More
3M™ Particulate Respirator, 8300 Series
Provide comfortable, effective respiratory protection against dust particles and/or non-volatile liquid particles. The 3M 8300 Series Particulate Respirator gives effective filtration with low breathing resistance for consistent high-quality performance.
Brand:  3M™ 7100010813
Code : NEW
Additional Details : Weight : 0.00100kg
Product Code. 11746003
  • £47.92 / Pack of 10
In Stock 2
Add to basket
You may only order up to the available stock. For more details call customer services on 01509 555 500
Description
Description
Provide comfortable, effective respiratory protection for use in industries where workers will be exposed to dust particles and/or non-volatile liquid particles.
  • Tested and certified to AS/NZS 1716:2003
  • Robust outer shell improves collapse resistance
  • M-Noseclip adjusts easily for fewer pressure points and greater comfort on the nose
  • Braided headbands improve comfort on the neck, face, and head with a secure feel
  • 3M™ Advanced Electret Filter Material gives effective filtration with low breathing resistance for consistent high-quality performance
  • The super-soft, cushioned inner lining helps workers stay comfortable
  • Models that include the Cool Flow™ exhalation valve offer improved comfort in hot humid environments and/or where work is hard and physical
  • Colored headbands for easy identification: yellow for P1, blue for P2
Specifications
Specifications
10 Pack
Respirator
FFP3
Elastic
Hull
EN149: 2001 A1: 2009, CE
Respirator
Hull Shaped
Protection against solid aerosols, watery and non-volatile liquids
Protection against solid aerosols, watery and non-volatile liquids
Yes
Videos