Learn More
HTMT dimaleate, Tocris Bioscience™
H1/ H2 agonist
Brand:  Tocris (Bio-Techne) 064650
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13487843
  • £492.72 / 50mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
H1 and H2 receptor agonist. 4x104 times more active than histamine in H2-mediated effects in natural suppressor cells. Increases intracellular Ca2+ and IP3 in lymphocytes through a binding site other than H1, H2 or H3.
Specifications
Chemical Identifiers
195867-54-0
618.607
6-[2-(4-Imidazolyl)ethylamino]-N-(4-trifluoromethylphenyl)heptanecarboxamide dimaleate
C27H37F3N4O9
KIMGHGOBUFFPNE-UHFFFAOYSA-N
77519427
Specifications
HTMT dimaleate
>99%
6-[2-(4-Imidazolyl)ethylamino]-N-(4-trifluoromethylphenyl)heptanecarboxamide dimaleate
CC(CCCCC(=O)NC1=CC=C(C=C1)C(F)(F)F)NCCC2=CN=CN2.C=CC(=O)O.C=CC(=O)O.C(=O)O.C(=O)O
618.607
614.57
195867-54-0
C27H37F3N4O9
KIMGHGOBUFFPNE-UHFFFAOYSA-N
formic acid;6-[2-(1H-imidazol-5-yl)ethylamino]-N-[4-(trifluoromethyl)phenyl]heptanamide;prop-2-enoic acid
77519427