missing translation for 'onlineSavingsMsg'
Learn More

HSP70, clone: 1B2hsp70, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9115-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15507586

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »

Disclaimers:

For Research Use Only.


Description

Description

The monoclonal antibody 1B2hsp70 recognizes HSP (heat shock protein) 70. Hsps are highly conserved molecular chaperones that associate with proteins to stabilize or aid in the folding of new proteins or denatured proteins following chemical or physical stress. Hsp expression is both constitutive and inducible depending on the cell and its surrounding conditions. Hsps have been known to be upregulated in conditions such as cancer, ischemic injury, inflammatory, and demyelinating disease.

This antibody recognizes the Hsp70 family member 1A found in the cytoplasm and nucleus of numerous cell types. Given the high degree of conservation of this protein across species, the 1B2hsp70 clone is expected to recognize canine and monkey HSP70 1a.
Specifications

Specifications

HSP70
1B2hsp70
Unconjugated
Purified
Murine
100μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HSPA1A
IgG1
RUO
Primary
Human
Videos