missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse HSF4 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | HSF4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
HSF4 Polyclonal specifically detects HSF4 in Mouse samples. It is validated for Western Blot.Specifications
HSF4 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
ATHSF4, AT-HSFB1, cataract, Marner, CTM, heat shock factor protein 4, heat shock transcription factor 4CTM, hHSF4, HSF 4, HSFB1, HSTF 4 | |
HSF4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9R0L1 | |
3299 | |
E. coli-derived recombinant mouse HSF4 Gly343-Pro492 Accession # Q9R0L1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse HSF4 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) HSF4 is observed and less than 5% cross-reactivity with recombinant mouse HSF1 and rhHSF2 is observed. |
For Research Use Only