Learn More
HSD17B14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554417
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ATGTRYAGKV VVVTGGGRGI GAGIVRAFVN SGARVVICDK DESGGRALEQ ELPGAVFILC DVTQEDDVKT LVSETIR Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 71%.
HSD17B14 Polyclonal antibody specifically detects HSD17B14 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
HSD17B14 | |
Polyclonal | |
Unconjugated | |
HSD17B14 | |
0610039E24Rik; 17-beta-HSD 14; 17-beta-hydroxysteroid dehydrogenase 14; 17-beta-hydroxysteroid dehydrogenase DHRS10; dehydrogenase/reductase (SDR family) member 10; dehydrogenase/reductase SDR family member 10; DHRS10; HSD17B14; hydroxysteroid (17-beta) dehydrogenase 14; hydroxysteroid 17-beta dehydrogenase 14; retinal short-chain dehydrogenase/reductase 3; retinal short-chain dehydrogenase/reductase retSDR3; retSDR3; SDR3; SDR47C1; Short chain dehydrogenase/reductase family 47C member 1; short chain dehydrogenase/reductase family 47C, member 1; testicular tissue protein Li 52; UNQ502/PRO474 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51171 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BPX1 | |
HSD17B14 | |
Recombinant protein corresponding to Human HSD17B14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only