Learn More
Duran™ HPLC Bottles
Meets USP Standard. DURAN™ HPLC Bottles allow safe transfer of liquid media within a closed and sterile system (evaporation is reduced).
£142.00 - £153.00
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10554493 View Documents Duran™
1129821
500 mL £142.00
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
15-06-2020
Add to basket
10014432 View Documents Duran™
1129820
1000 mL £153.00
Pack of 2 View alternative products
Save Up to 
»
Estimated Shipment
15-06-2020
Add to basket
Description
Description
  • Easy-to-read scale and large labelling field in fired-on, highly durable white ceramic
  • Complete system comprising: DURAN™ pressure plus laboratory bottle with 4-port screw cap (PP); four screw connections (black, M8 thread) and silicone seals
  • Connection of different hose diameters (1.6 mm and 3.2 mm) as well as sterile pressure equalisation sets (membrane filter 0.2 μm)
  • Unused ports can be sealed with silicone blanking seals