missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564441
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ARAYSQSDPK QPPSGTALKQ PAVVYPWMKK V Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.Specifications
HOXD4 | |
Polyclonal | |
Unconjugated | |
HOXD4 | |
6030436D05Rik; HHO.C13; homeo box D4; homeobox D4; homeobox protein HHO.C13; Homeobox protein Hox-4.2; homeobox protein Hox-4B; Homeobox protein Hox-5.1; homeobox protein Hox-D4; HOX4; Hox-4.2; Hox-4.2, mouse, homolog of homeo box X; HOX4B; Hox-5.1; HOXD4; Hoxd-4; LOW QUALITY PROTEIN: homeobox protein Hox-D4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3233 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09016 | |
HOXD4 | |
Recombinant protein corresponding to Human HOXD4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only