missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565827
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: PNTKVRSAPPA GAAPSTLSAA TPGTSEDHSQ SATPPEQQRA EDITRL Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
HOXC4 Polyclonal antibody specifically detects HOXC4 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
HOXC4 | |
Polyclonal | |
Unconjugated | |
HOXC4 | |
cp19; homeo box 3E; homeo box C4; homeobox C4; Homeobox gene C4; homeobox protein CP19; Homeobox protein Hox-3.5; homeobox protein Hox-3E; Homeobox protein Hox-C4; homeobox protein R3; HOX3; Hox-3.5; Hox-35; HOX3E; Hox3r3; Hoxc4; Hoxc-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3221 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09017 | |
HOXC4 | |
Recombinant Human HOXC4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only