missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567398
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: NNKDKFPSSKC EQEELEKQKL ERAPEAADEG DAQK Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.
This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in development. Increased expression of this gene is associated with colorectal cancer. Mice that have had the murine ortholog of this gene knocked out exhibit an excessive pathologic grooming behavior. This behavior is similar to the behavior of humans suffering from the obsessive-compulsive spectrum disorder trichotillomania.Specifications
HOXB8 | |
Polyclonal | |
Unconjugated | |
HOXB8 | |
homeo box 2D; homeo box B8; homeobox B8; homeobox gene B8; homeobox protein Hox-2.4; Homeobox protein Hox-2D; homeobox protein Hox-B8; homeobox protein R1a; HOX2; Hox-2.4; Hox2.4 homeobox homolog; HOX2D; Hox2r1a; Hoxb8; Hoxb-8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P17481 | |
HOXB8 | |
Recombinant Human HOXB8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only