missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568211
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse and Rat samples.
HOXB3 Polyclonal antibody specifically detects HOXB3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HOXB3 | |
Polyclonal | |
Unconjugated | |
HOXB3 | |
homeo box 2G; homeo box B3; homeobox B3; homeobox protein Hox-2.7; Homeobox protein Hox-2G; homeobox protein Hox-B3; Homeobox protein MH-23; HOX2; Hox-2.7; HOX2G; HOXB3; Hoxb-3 | |
Rabbit | |
affinity chromatography | |
RUO | |
15410, 303488, 3213 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P09026, P14651 | |
HOXB3 | |
Synthesized peptide derived from human HOXB3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |