missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550510
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total HOXA4 protein.
HOXA4 Polyclonal antibody specifically detects HOXA4 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
HOXA4 | |
Polyclonal | |
Unconjugated | |
HOXA4 | |
AV206827; Dfd-like protein; homeo box A4; homeobox A4; homeobox protein Hox-1.4; Homeobox protein Hox-1D; homeobox protein Hox-A4; homeobox protein Hox-A4-like; homeobox protein MH-3; HOX1; Hox-1.4; Hox-1.4-like protein; Hox-14; HOX1D; Hoxa4; Hoxa-4; LOC100912525 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3201 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q00056 | |
HOXA4 | |
Synthetic peptide corresponding to a region derived from internal residues of human homeobox A4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |