Learn More
Miele™ Housing unit, Aqua Purificator, stainless steel, for PG 8595 laboratory glass washers, Miele
Brand:  Miele™ 10087320
Code : NEW
Additional Details : Weight : 30.00000kg
Product Code. 15407724
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Housing unit for two water demineralisation cartridges VEP2000/VEP2800
  • Compatible with PG 8583, PG 8593, PG 8583 CD Miele glass laboratory washers
  • Generally recommended quality for final rinse < 19μS/cm
  • H 835 (820), W 300, D 600 mm
  • Freestanding unit, can be built under
  • Outer panelling in stainless steel\white