missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Hostess™ Toilet Tissue Rolls

Dissolves in cold water so it won’t block drains. Kimberly-Clark™ HOSTESS™ Small Toilet Tissue Rolls are manufactured from 100% recycled materials and are fully biodegradable.

Brand:  Kimberly-Clark™ 8641

Code : 02

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12744246

  • £45.50 / 36 rolls


Estimated Shipment
10-05-2018
Add to basket

Description

Description

  • White perforated toilet tissue
  • Suppliled in packs of 36.
  • 100% recycled
  • Fully biodegradable to reduce the risk of blocked toilets
  • ISO 9001 and ISO 14001and EU Ecolabel certified
Specifications

Specifications

ISO 9001 and ISO 14001, EU Ecolabel certified
11.5 x 9.4 cm
18 x 2 rolls x 650 sheets
1
Ideal for small, office and hotel washrooms, especially where economy is important
Small Roll Toilet Tissue
White
Small Roll
1-ply
9.4 cm
11.5 cm
Videos