missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ HOSTESS™ Tela Fold Hand Towels

Self-presenting, dispensed open and ready to use. Kimberly-Clark™ HOSTESS™ Telafold Hand Towels include a break every seven sheets to stop streaming, helping to reduce costs and waste.

Brand:  Kimberly-Clark™ 6876

Code : 02

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12734316

  • £70.00 / Pack of 12


Estimated Shipment
16-02-2018
Add to basket

Description

Description

  • Small, 1-ply hand towels
  • Self-presenting, dispensed open and ready to use
  • Break every seven sheets stops streaming and helps to reduce costs and waste
  • Ideal for busy washrooms -- in schools and other public facilities where economy is important.
  • Carries the EU Ecolabel
Specifications

Specifications

EU Ecolabel
24 x 23 cm
12 x 224 sheets
23 cm
24 cm
Blue
Tela Fold
1-ply
Ideal for busy washrooms, for example in schools and other public facilities where economy is important
Small Folded Hand Towel
Videos