missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Hostess™ Hand Towels

Brand:  Kimberly-Clark™ 6805

Code : 02

Additional Details : Weight : 3.50000kg

Product Code. 12704316

  • £87.05 / Pack of 3328


Estimated Shipment
02-03-2018
Add to basket

Specifications

Specifications

EU Ecolabel
33 x 22.5 cm
3328/Pk.
1
Ideal for use in busy public venues and work environment washrooms
Paper
White
C-fold
1-Ply
22.5 cm
33 cm
Hand Towel
Videos