missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Screw Cap Open Top for Tube Connection
Click to view available options
Blue
Description
Flexible GLS 80™ connection system for the use of semi-rigid hoses.
Flexible GLS 80™ cap connection system for use with semi-rigid tubing, suitable for DURAN™ GLS 80™ laboratory and media bottles. Five different tube diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation through membrane liter possible. Unused ports can be provided with a blind cap. Exemplary applications: Safe transfer of liquid media within a closed and sterile system.
- The flexible modular system allows individual combination of the various components according to requirements and application
- Five different hose diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected
- NS 29/32 connection in the middle for tight closure
- Sterile pressure equalisation through membrane filter possible
- Unused ports can be fitted with a GL 18 blind cap
- Compatible with all GLS 80 threads and therefore versatile
Specifications
Closure Material | Polypropylene, PTFE |
Closure Type | Screw Cap |
Diameter (Metric) | 87 mm |
Product Type | Screw Cap Open Top |
Color | Blue |
Description | Screw cap for tube connection, blue, GL 18 |
For Use With (Application) | Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments; Photo-bioreactor |
Material | Polypropylene, PTFE |
Quantity | 2/Pk. |
Sterility | Sterile |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'