Learn More
HORMAD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565021
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: DKDVEDEQEHY TSDDLDIETK MEEQEKNPAS SELEEPSLVC EEDEIMRSKE SPDLSISHSQ VEQLVNK Highest antigen sequence identity to the following orthologs - mouse 50%, rat 47%.
HORMAD1 Polyclonal antibody specifically detects HORMAD1 in Human samples. It is validated for Immunocytochemistry.Specifications
HORMAD1 | |
Polyclonal | |
Unconjugated | |
HORMAD1 | |
4921522K05Rik; Cancer/testis antigen 46; CT46; HORM1; HORMA domain containing 1; HORMA domain containing protein; HORMA domain-containing protein 1; HORMAD1; Newborn ovary HORMA protein; Nohma; RGD1564960; testis tissue sperm-binding protein Li 86P | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84072 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86X24 | |
HORMAD1 | |
Recombinant Human HORMAD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only