missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hopx Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549449
Additional Details : Weight : 0.01000kg
Description
Hopx Polyclonal antibody specifically detects Hopx in Human, Mouse samples. It is validated for Western Blot.Specifications
Hopx | |
Polyclonal | |
Unconjugated | |
Hopx | |
1110018K11Rik; 1200015P04Rik; 2300002F06Rik; AI848177; AW490897; Cameo; GIIg15b; global ischemia induced protein GIIG15B; global ischemia-induced gene 15B protein; global ischemia-induced protein 15B; Hdop; HOD; homeobox only domain; homeobox-only protein; homeodomain only protein; homeodomain-only protein; Hop; HOP homeobox; Hopx; LAGY; Lung cancer-associated Y protein; mOB1; NECC1; not expressed in choriocarcinoma clone 1; Not expressed in choriocarcinoma protein 1; OB1; Obl; odd homeobox 1; odd homeobox protein 1; SMAP31; Toto | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
74318, 84525 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide | |
Q8R1H0, Q9BPY8 | |
Hopx | |
KLH conjugated synthetic peptide between 22-54 amino acids from the Central region of Mouse Hopx. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |