missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homophthalic anhydride, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It is used as a chemical and organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a chemical and organic intermediates.
Solubility
Decomposes in water.
Notes
Moisture sensitive. Storage temperature 15 - 30°C. Store under inert gas. Store away from exposure to moisture, strong oxidizing agents, strong acids, strong bases.
It is used as a chemical and organic intermediates.
Solubility
Decomposes in water.
Notes
Moisture sensitive. Storage temperature 15 - 30°C. Store under inert gas. Store away from exposure to moisture, strong oxidizing agents, strong acids, strong bases.
Specifications
Melting Point | 140°C to 144°C |
Quantity | 10 g |
Beilstein | 5244 |
Sensitivity | Moisture sensitive |
Solubility Information | Decomposes in water. |
Formula Weight | 162.14 |
Percent Purity | ≥97% |
Chemical Name or Material | Homophthalic anhydride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'