missing translation for 'onlineSavingsMsg'
Learn More

Holmium rod, 6.35mm (0.25in) dia, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7059139
162.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 mm
50 mm
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-60-0
Ho
164.93
MFCD00011049
KJZYNXUDTRRSPN-UHFFFAOYSA-N
atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
23988
CHEBI:49648
holmium
[Ho]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-60-0
Ho
164.93
MFCD00011049
KJZYNXUDTRRSPN-UHFFFAOYSA-N
atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
23988
CHEBI:49648
holmium
[Ho]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in mylar under argon
TRUSTED_SUSTAINABILITY
Quantity 10 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.28g/mm
Form Cast Rod
Assay (metals basis)
Chemical Name or Material Holmium rod, ≈6.35mm (0.25 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'