missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Holmium, plasma standard solution, Specpure™, Ho 10μg/mL

Brand:  Alfa Aesar™ 045255.AE

Code : NEW

Additional Details : Weight : 0.00010kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 100mL

Product Code. 15446855

  • £110.30 / 100mL


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Specifications

Holmium, plasma standard solution
Specpure™
Ho2O3 in 2% HNO3
UN3264
Ho 10μg/mL
Liquid
MFCD00011049
100mL
Videos
SDS