missing translation for 'onlineSavingsMsg'
Learn More

Holmium Perchlorate EU Pharmacopoeia, Fisher Chemical™

Catalog Number 12970424
Click to view available options
Quantity:
125 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Concentration 100mg/mL
Packaging Bottle
Quantity 125 mL
Analyte or Component Names Hf, Ir, Sb, Sn, Ta, Ti, Zr
Solvent or Matrix HCl 15%; HF, HNO3
compliance-icons compliance-icons
Product Identifier
  • Holmium perchlorate solution. European Pharmacopoeia Ref: 1043101 (RP 089)
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'