missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Holmium(III) phosphate hydrate, 99.9% (REO)

Brand:  Alfa Aesar™ 041994.04

Code : NEW

Additional Details : Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11354997

  • £27.10 / 2g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Holmium(III) phosphate hydrate
259.87 (Anhydrous)
MFCD00467736

Specifications

Holmium(III) phosphate hydrate
99.9%
259.87 (Anhydrous)
2g
Powder
(REO)
MFCD00467736
Videos
SDS