missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7049519
54.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14483-18-2
H10HoN3O14
441.017
MFCD00149737
BJUGWWDCFYEYOA-UHFFFAOYSA-N
holmium nitrate pentahydrate, ho.3no3.5h2o, holmium iii nitrate hydrate, holmium nitrate, holmium nitrate-water 1/3/5, holmium 3+ pentahydrate trinitronate, holmium iii nitrate pentahydrate, holmium 3+ pentahydrate trinitrate, nitric acid,holmium 3+ salt, pentahydrate 8ci,9ci, holmium iii nitrate pentahydrate trace metals basis
25021754
holmium(3+);trinitrate;pentahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14483-18-2
H10HoN3O14
441.017
MFCD00149737
BJUGWWDCFYEYOA-UHFFFAOYSA-N
holmium nitrate pentahydrate, ho.3no3.5h2o, holmium iii nitrate hydrate, holmium nitrate, holmium nitrate-water 1/3/5, holmium 3+ pentahydrate trinitronate, holmium iii nitrate pentahydrate, holmium 3+ pentahydrate trinitrate, nitric acid,holmium 3+ salt, pentahydrate 8ci,9ci, holmium iii nitrate pentahydrate trace metals basis
25021754
holmium(3+);trinitrate;pentahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3]

Holmium(III) nitrate pentahydrate is used to prepare holmium-beta-cyclodextrin complex by reacting with beta- cyclodextrin, which posses potential pharmacological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline Aggregates
Quantity 10 g
UN Number UN1477
Solubility Information Soluble in water.
Formula Weight 441.02 (350.94 Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Holmium(III) nitrate pentahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Holmium(III) nitrate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P380+P375-In case of fire: Evacuate area. Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid, holmium(III) pentahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'