missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 15432107
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
13760-78-6
F3Ho
221.93
MFCD00016142
FDIFPFNHNADKFC-UHFFFAOYSA-K
holmium(3+) trifluoride
[F-].[F-].[F-].[Ho+3]
This item is not returnable. View return policy
13760-78-6
F3Ho
221.93
MFCD00016142
FDIFPFNHNADKFC-UHFFFAOYSA-K
holmium(3+) trifluoride
[F-].[F-].[F-].[Ho+3]

Holmium(III) fluoride, anhydrous is used to produce the strongest artificially- generated magnetic fields, nuclear control rods, yellow-colored glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1143°C
Physical Form Powder
Quantity 2 g
UN Number UN3288
Solubility Information Insoluble in water.
Formula Weight 221.93
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.6 g/mL
Assay (REO)
Chemical Name or Material Holmium (III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'