missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) bromide, ultra dry, 99.98% (REO), Thermo Scientific™

Catalog Number p-7049414
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
13825-76-8
MFCD00049744
This item is not returnable. View return policy
13825-76-8
MFCD00049744

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 914°C
Physical Form Powder
Quantity 1 g
Merck Index 14,4727
Formula Weight 404.66
Percent Purity 99.98%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Holmium(III) bromide, Ultra dry
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'