missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7049521
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
312619-49-1
C6H9HoO6
342.06
MFCD00150648
HWQRVFRWLQGBFV-UHFFFAOYSA-K
holmium iii acetate hexahydrate, holmium iii acetate hydrate, reacton reo, holmium iii acetatehexahydrate, holmium acetate-water 1/3/6
holmium(3+) triacetate
[Ho+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
This item is not returnable. View return policy
312619-49-1
C6H9HoO6
342.06
MFCD00150648
HWQRVFRWLQGBFV-UHFFFAOYSA-K
holmium iii acetate hexahydrate, holmium iii acetate hydrate, reacton reo, holmium iii acetatehexahydrate, holmium acetate-water 1/3/6
holmium(3+) triacetate
[Ho+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Holmium(III) acetate hydrate is used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline Aggregates
Quantity 2 g
Solubility Information Soluble in water.
Formula Weight 342.07 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Assay (REO)
Chemical Name or Material Holmium(III) acetate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'