Learn More
Hoefer™ TE70X and TE77X Semidry Blotting Systems
Perform uniform transfers of proteins and nucleic acids from polyacrylamide and agarose gels in less than one hour
Brand:  Hoefer™ TE70X
Code : 04
Additional Details : Weight : 3.70000kg
 View more versions of this product
Product Code. 11985338
  • £1095.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Transfer up to two polyacrylamide gels in a stack
  • Use minimal buffer
  • Do not generate the excessive heat that can distort bands or denature protein
  • Utilize durable iridium oxide and stainless-steel electrodes to assure uniform, contamination-free transfers
  • Offer automatic transfer-stopping feature to prevent overheating when buffer is depleted
  • Available with built-in power supply to save space and simplify transfer
Specifications
Specifications
Semi Dry Transfer Unit
48cm
Electrophoresis blotting system
Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (14 x 16cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™ masks (16.5 x
Up to 80%
45°C
15w
10cm
38cm
18.89 x 15 x 4 in. (48 x 38 x 10cm)
500mA
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
30V
Videos
Documentation
Technical Documents
View accessory products