Learn More
Hoefer™ TE70X and TE77X Semidry Blotting Systems
Perform uniform transfers of proteins and nucleic acids from polyacrylamide and agarose gels in less than one hour
Brand:  Hoefer™ TE77XP
Code : 04
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11915348
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Includes: Molded base with iridium oxide anode, top assembly with stainless-steel cathode, power cables, 25 sheets of blotting paper, 50 sheets of cellophane (Mfr. Nos. TE70X and TE70XP only), two solid Mylar™ masks
Description
Description
  • Transfer up to two polyacrylamide gels in a stack
  • Use minimal buffer
  • Do not generate the excessive heat that can distort bands or denature protein
  • Utilize durable iridium oxide and stainless-steel electrodes to assure uniform, contamination-free transfers
  • Offer automatic transfer-stopping feature to prevent overheating when buffer is depleted
  • Available with built-in power supply to save space and simplify transfer
Certifications
CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1
Specifications
Specifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Semi Dry Transfer Unit
Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™
45°C
18.89 in.
15 in.
4 in.
30 V
500 mA
Electrophoresis Blotting System
Up to 80%
113°F
18.89 x 15 x 4 in. (48 x 38 x 10 cm)
48 cm
38 cm
10 cm
15 w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products