missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SUB15 Standard Submarine Combs

Accessories for SUB15 Standard Submarine

Brand:  Hoefer™ SUB15-C2-30MC

UNSPSC : 41105305

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 12665770

  • £60.55 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

0.2cm
Comb
30
0.25cm
0.5cm
SUB15 standard submarine
24μL