Learn More
Hoefer™ SUB15 Standard Submarine Combs
Accessories for SUB15 Standard Submarine
Brand:  Hoefer™ SUB15-C2-20
Code : 04
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12655770
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Ranges from 10, 16, 20, 25 and 30 Wells
  • 1, 1.5 and 2 mm Thickness
  • 2.5 to 12.8 mm Well Width
  • Min. 12, max. 128 μL Sample Volume
Specifications
Specifications
SUB15 Standard Submarine
20
0.2 cm
0.23 in.
0.19 in.
Comb
0.07 in.
58 μL
0.59 cm
0.5 cm