missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SUB15 Standard Submarine Combs

Accessories for SUB15 Standard Submarine

Brand:  Hoefer™ SUB15-C1-10

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12645756

  • £49.68 / Each


Estimated Shipment
16-04-2021
Add to basket

Description

Description

Specifications

Specifications

0.5 cm
Comb
0.1 cm
1.28 cm
SUB15 Standard Submarine
10
64 μL
Videos
SDS
Documents