missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Spacers for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units

Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units

Brand:  Hoefer™ SE6119-2-1.0

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 12034657

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

16cm
2cm
Spacer
2 x 16cm x 1.0mm
0.1cm
SE 600 Chroma, SE 600 standard, SE 400 vertical electrophoresis unit
2/Pk.
Videos
Documentation

Technical Documents